missing translation for 'onlineSavingsMsg'
Learn More

Safetec™ Red-Z™ Biohazard Fluid Control Powders

Catalog No. 19306694
Encompass
Change view
Click to view available options
Type:
Green Z Spill Control Solidifier
Red Z Spill Control Solidifier
Packaging Quantity:
5 oz.
11 oz.
15 oz.
3.5 lb.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
19306694 Red Z Spill Control Solidifier 11 oz.
19023901 Green Z Spill Control Solidifier 15 oz.
19306693 Red Z Spill Control Solidifier 5 oz.
19290179 Red Z Spill Control Solidifier 3.5 lb.
19023899 Green Z Spill Control Solidifier 5 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19306694 Supplier Safetec™ Supplier No. SF41102

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For safe cleanup of spills of bodily fluids

Total fluid control encapsulators solidify and deodorize blood, vomit, urine or other potentially hazardous, water-based fluids. 

  • Red-Z solidifies and contains chlorine to deodorize potentially infectious blood and bodily fluids
  • Green-Z solidifies and can be used in the cleanup of blood, bodily fluids, chemotherapy, glutaraldehyde, and mercury spills

 

Order Info

Supplied in shaker-top bottles.

Specifications

Certifications/Compliance OSHA
Type Red Z Spill Control Solidifier
Format Powder
For Use With (Application) Solidify and deodorize potentially Infectious blood and body fluids
Packaging Quantity 11 oz.
Weight (Metric) 0.31 kg