Learn More
Plasticoid Red Rubber Sleeve Stoppers for Serum and Vaccine Bottles
Supplier:  Plasticoid 20230
 View more versions of this product
Catalog No. 14-126DD
Only null left
Explore available promotions
Description
Description
  • Comprised of dry natural rubber
  • Folds down over neck of bottle
  • With hollow plugs, rounded corners and needle guide
  • Easily perforated with hypodermic needles
  • Self-sealing
Specifications
Specifications
Red
19
Rubber
Serum and vaccine bottles
Sleeve Stopper
15
1.9cm
1.58cm
Product Suggestions
Product Suggestions