Learn More
United Scientific™ Rectangular Block
Description
- Rectangular blocks are available in a variety of sizes and are designed for use as an index of refraction plate.
- Available in Glass or Acrylic.
- Available in five variations.
Specifications
Specifications
| Material | Acrylic |
| Product Type | Prism |
| Shape | Rectangular |
| Dimensions (L x W x H) | 115 x 65 x 20 mm |
Warning: Cancer and reproductive harm