Learn More
United Scientific™ Rectangular Block
Ideal for use demonstrating index of refraction.
Supplier: United Scientific™ RCB100
Description
- Rectangular blocks are available in a variety of sizes and are designed for use as an index of refraction plate.
- Available in Glass or Acrylic.
- Available in five variations.
Chemical Identifiers
| Prism | |
| Rectangular |
Specifications
| Acrylic | |
| Prism | |
| Rectangular | |
| 100 x 75 x 20 mm |
Warning: Cancer and reproductive harm