Recombinant Rubella Virus spike glycoprotein E1 protein (Fc Chimera) is a Rubella virus strain Therien Fragment protein, in the 583 to 1025 aa range, expressed in HEK 293, with >=95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Structural polyprotein, p110
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement