Learn More
ACROBiosystems Inc Protein A-coupled Magnetic Beads (recommended for MPCLIA)
Catalog No. 502332062
Encompass
The Protein A-coupled Magnetic Beads are 2.8 m superparamagnetic particles covalently coupled to a highly pure form of protein A. The beads can be used to capture the antibodies in Chemiluminescence procedures.The Protein A is a 42 kDa surface protein, It binds the heavy chain within the Fc region of most immunoglobulins and also within the Fab region in the case of the human VH3 family.The Anti-Protein A-coupled Magnetic Beads is easy to capture the antibodies, and the bounded antibody have no activity lost, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.
Catalog No. 502332062 Supplier ACROBiosystems Inc Supplier No. MPCA001100MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Protein A-coupled Magnetic Beads are 2.8 m superparamagnetic particles covalently coupled to a highly pure form of protein A. The beads can be used to capture the antibodies in Chemiluminescence procedures.The Protein A is a 42 kDa surface protein, It binds the heavy chain within the Fc region of most immunoglobulins and also within the Fab region in the case of the human VH3 family.The Anti-Protein A-coupled Magnetic Beads is easy to capture the antibodies, and the bounded antibody have no activity lost, this ready to use products could greatly save your protein coupling time and hassle, and help us get the best performance and highly reproducible results.