Learn More
ACROBiosystems Inc FITC-Labeled Human Siglec-3 / CD33 Protein, His Tag
Catalog No. 502019657
Encompass
Recombinant Protein;200UG;FITC-Labeled Human Siglec-3, His Tag (CD3-HF224) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). It is the FITC labeled form of Human Siglec-3, His Tag (CD3-H5226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 g/mL (100 L/well) can bind FITC-Labeled Human Siglec-3, His Tag (Cat. No. CD3-HF224) with a linear range of 0.02-0.625 g/mL (QC tested).
Catalog No. 502019657 Supplier ACROBiosystems Inc Supplier No. CD3HF224200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;FITC-Labeled Human Siglec-3, His Tag (CD3-HF224) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1). It is the FITC labeled form of Human Siglec-3, His Tag (CD3-H5226).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.6 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Siglec-3 Antibody at 1 g/mL (100 L/well) can bind FITC-Labeled Human Siglec-3, His Tag (Cat. No. CD3-HF224) with a linear range of 0.02-0.625 g/mL (QC tested).