Learn More
ACROBiosystems Inc Biotinylated Human uPAR / PLAUR Protein, His,Avitag™
Catalog No. 502019856
Encompass
Recombinant Protein;200UG;Biotinylated Human uPAR, His,Avitag (UPR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 35.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PLAU, His Tag (Cat. No. PLU-H5229) at 5 g/mL (100 L/well) can bind Biotinylated Human uPAR, His,Avitag (Cat. No. UPR-H82E7) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 502019856 Supplier ACROBiosystems Inc Supplier No. UPRH82E7200UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;200UG;Biotinylated Human uPAR, His,Avitag (UPR-H82E7) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 35.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PLAU, His Tag (Cat. No. PLU-H5229) at 5 g/mL (100 L/well) can bind Biotinylated Human uPAR, His,Avitag (Cat. No. UPR-H82E7) with a linear range of 0.3-2 ng/mL (QC tested).