Learn More
ACROBiosystems Inc Human NKG2C / CD159c Protein, His Tag
Catalog No. 502019438
Encompass
Recombinant Protein;100UG;Human NKG2C, His Tag (NKC-H5248) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (Accession # P26717-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa.
Catalog No. 502019438 Supplier ACROBiosystems Inc Supplier No. NKCH5248100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Human NKG2C, His Tag (NKC-H5248) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (Accession # P26717-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.7 kDa.