Learn More
ACROBiosystems Inc Cynomolgus FOLR2 Protein, His Tag
Catalog No. 502019493
Encompass
Recombinant Protein;100UG;Cynomolgus FOLR2, His Tag (FO2-C52H8) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - His 245 (Accession # A0A2K5U027-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 502019493 Supplier ACROBiosystems Inc Supplier No. FO2C52H8100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Cynomolgus FOLR2, His Tag (FO2-C52H8) is expressed from human 293 cells (HEK293). It contains AA Thr 34 - His 245 (Accession # A0A2K5U027-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-43 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.