Learn More
Abcam Inc Recombinant Influenza A Influenza A Virus Nucleoprotein (His tag), 100UG
Catalog No. 502587374
Encompass
Recombinant Influenza A Influenza A Virus Nucleoprotein (His tag) is a Influenza A virus (A/Beijing/353/1989(H3N2)) Full Length protein, in the 1 to 498 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: Nucleoprotein, Nucleocapsid protein, Protein N, NP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502587374 Supplier Abcam Inc Supplier No. AB224859100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Influenza A Influenza A Virus Nucleoprotein (His tag) is a Influenza A virus (A/Beijing/353/1989(H3N2)) Full Length protein, in the 1 to 498 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: Nucleoprotein, Nucleocapsid protein, Protein N, NP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement