missing translation for 'onlineSavingsMsg'
Learn More

Abcam Inc Recombinant Human ErbB2 / HER2 protein, 1MG

Catalog No. 502704074
Encompass

Recombinant Human ErbB2 / HER2 protein is a Human Fragment protein, in the 23 to 652 aa range and expressed in Mammalian.

Protein/ Peptide/ Enzyme Synonyms: CD340, HER2, MLN19, NEU, NGL, ERBB2, Receptor tyrosine-protein kinase erbB-2, Metastatic lymph node gene 19 protein, Proto-oncogene Neu, Proto-oncogene c-ErbB-2, Tyrosine kinase-type cell surface receptor HER2, p185erbB2, MLN 19

Conjugation: Unconjugated

The product is subject to the following: Abcam Restricted Use Statement

Catalog No. 502704074 Supplier Abcam Inc Supplier No. AB2705861MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Human ErbB2 / HER2 protein is a Human Fragment protein, in the 23 to 652 aa range and expressed in Mammalian.

Protein/ Peptide/ Enzyme Synonyms: CD340, HER2, MLN19, NEU, NGL, ERBB2, Receptor tyrosine-protein kinase erbB-2, Metastatic lymph node gene 19 protein, Proto-oncogene Neu, Proto-oncogene c-ErbB-2, Tyrosine kinase-type cell surface receptor HER2, p185erbB2, MLN 19

Conjugation: Unconjugated

The product is subject to the following: Abcam Restricted Use Statement