Learn More
Abcam Inc Recombinant Hepatitis C Virus Core Antigen protein (FITC), 100UG
Catalog No. 502710695
Encompass
Recombinant Hepatitis C Virus Core Antigen protein (FITC) is a Hepatitis C virus (isolate 1) protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Genome polyprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502710695 Supplier Abcam Inc Supplier No. AB49021100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Hepatitis C Virus Core Antigen protein (FITC) is a Hepatitis C virus (isolate 1) protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: Genome polyprotein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement