Recombinant Hepatitis B Virus Core Antigen protein is a Hepatitis B virus ayw/France/Tiollais/1979 Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, ELISA.
Protein/ Peptide/ Enzyme Synonyms: Capsid protein, Core antigen, Core protein, HBcAg, p21.5, C
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement