Learn More
Abcam Inc Recombinant E. coli Carbonic anhydrase 2/CA2 protein, 20UG
Catalog No. 502676822
Encompass
Recombinant E. coli Carbonic anhydrase 2/CA2 protein is a Escherichia coli K-12 Full Length protein, in the 1 to 220 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: cynT2, yadF, b0126, JW0122, can, Carbonic anhydrase 2, Carbonate dehydratase 2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502676822 Supplier Abcam Inc Supplier No. AB20830920UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant E. coli Carbonic anhydrase 2/CA2 protein is a Escherichia coli K-12 Full Length protein, in the 1 to 220 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: cynT2, yadF, b0126, JW0122, can, Carbonic anhydrase 2, Carbonate dehydratase 2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement