missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Recirculating Chillers

Catalog No. 05000002
Change view
Click to view available options
Cooling Capacity:
1400 W (at 15°C)
550 W (at 15°C)
900 W (at 15°C)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cooling Capacity
05000002 1400 W (at 15°C)
05000000 550 W (at 15°C)
05000001 900 W (at 15°C)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05000002 Supplier BUCHI Supplier No. 11F31402

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BUCHI Recirculating Chillers are easy to use and designed for use with Rotavapors, SyncorePlus, Multivapor, Kjeldahl systems, Extraction systems, and other suppliers' similar products.

  • Efficient distillation due to complete integration into the BUCHI rotary evaporation system
  • Temperature lock and temperature control in conjunction with the Interface I-300 / I-300 Pro.
  • Digital Display: Set and actual temperature
  • Saves time thanks to instant start with automatic and dynamic pressure adjustment without waiting until chiller reaches its set temperature
  • ECO-mode: Saves energy and reduces heat emission by switching into a standby mode when not in use
  • No water consumption
  • Maximizes distillation capacity while reducing solvent emissions due to intelligent integration of all process parameters using the Interface I-300 / I-300 Pro
  • Seamless plug and play integration into:
    • Fully automated BUCHI Rotavapor R-300, R-220 Pro systems including all process parameters
    • BUCHI parallel evaporators, extraction and Kjeldahl solutions

Specifications

Cooling Capacity 1400 W (at 15°C)
For Use With (Equipment) Up to 4 Rotavapor R-300
Type Recirculating Chiller
Model F-314