missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rec Protein A-HRP

Catalog No. 101123
Encompass
Change view
Click to view available options
Quantity:
3 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
101123 3 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 101123 Supplier Invitrogen™ Supplier No. 101123
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rec Protein A-HRP

This recombinant protein A is purified from cytosol of an E. coli strain expressing the genes coding for Staphylococcus aureus protein A. This version is conjugated with highly purified horseradish peroxidase.

Specifications

Conjugate HRP
Quantity 3 mg
Form Solution
Protein Subtype Protein A
Content And Storage 2-8°C. Do Not Freeze.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.