Learn More
Mopec Rear/Center, Standing Height Filler, Adjustable
Catalog No. 2123668
Change view
Click to view available options
Product Type:
Rear and Center Filler
Catalog No. Product Type
2123668 Rear and Center Filler
1 options
Catalog No. 2123668 Supplier Mopec Supplier No. LCFRHADJS
Only null left
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • Includes 1 standing height base filler, adjustable
  • The standing height rear/center filler is meant to be cut to length
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LCFRH-ADJS: 18.0 in. L x 1.0 in. W x 34.6 in. H
Performance
  • Fillers intended to create a seamless and polished look
  • Ensures that the cabinetry is well-integrated with the overall design of the room
Specifications
Product Type Rear and Center Filler