Learn More
Mopec Rear/Center Filler, Sitting Height
Description
Construction
- (1) Rear/Center Filler
Materials
- 304 stainless steel construction with a polished No. 4 finish
Dimensions
- LCFRL-07S: 7.0 in. L x 1.0 in. W x 27.1 in. H
Specifications
Specifications
| Product Type | Rear and Center Filler |