Learn More
Mopec Rear/Center Filler, ADA Height
Description
Construction
- Includes 1 ADA height rear/center filler
Materials
- 304 stainless steel construction with a polished No. 4 finish
Dimensions
- LCFRA-07S: 7.0 in. L x 1.0 in. W x 32.1 in. H
Performance
- Fillers help create a polished look by providing a finished appearance
- Ensures that cabinetry appears well-integrated into the overall design of the room
Specifications
Specifications
| Product Type | Rear and Center Filler |