missing translation for 'onlineSavingsMsg'
Learn More

Promega RealTime-Glo™ MT Cell Viability Assay

Catalog No. p-7068008
Encompass
Change view
Click to view available options
No. of Reactions:
10 x 100 reactions
100 reactions
1000 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRG9712 10 x 100 reactions
PRG9711 100 reactions
PRG9713 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG9712 Supplier Promega Supplier No. G9712

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The RealTime-Glo™ MT Cell Viability Assay is a non-lytic, homogeneous, bioluminescent method to determine in real time the number of viable cells in culture by measuring the reducing potential of cells and thus metabolism (MT).

  • Monitor Cell Viability in Real Time
  • Save time, cell culture and reagent costs
  • Rapidly detect changes in cell viability

Specifications

Content And Storage -20°C. Avoid prolonged exposure to light. Avoid multiple freeze-thaw cycles.
No. of Reactions 10 x 100 reactions