missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Real Time Electrophoresis Tank System

Run DNA gels, view separations as they occur, and capture gel images using a smart phone

$2,003.53

Specifications

Buffer 0.5 x or 1 x TAE or TBE
Certifications/Compliance CE
Detection Method Smart phone
For Use With (Equipment) Standard Electrophoresis Power Supply
Gel Compatibility Make gels using included casting set
View More Specs

Products 1
Catalog Number Mfr. No. Voltage Plug Type Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Plug Type Price Quantity & Availability  
14955205
View Documents Promotion Details
Fisherbrand™
E1200F
100/115 V US
Each for $2,003.53
Only null left
 
Description

Description

  • Tool for small DNA gel runs
  • Connects to any standard electrophoresis power supply
  • Compatible with SYBR, Gel green, SmartGlow, and other green fluorescent DNA stains
  • Imaging enclosure and photo filter allo gel imaging with any smart phone camera
  • Includes:
    • Gel running tank
    • Amber filter lid
    • Small blue LED transilluminator
    • Gel casting equipment (for casting your own gels)
      • Stand
      • Trays
      • Combs
  • Specifications

    Specifications

    0.5 x or 1 x TAE or TBE
    Smart phone
    Make gels using included casting set
    10.5 x 10 cm, 10.5 x 6 cm
    Agarose
    Gel tank, Blue LED illuminator, Filter cover, Gel casting set, Smart phone imaging enclosure
    Molded polycarbonate
    1
    1 Ea.
    10 to 20 μL
    12 or 22
    230 mL
    10 to 20 μL
    2 years
    7.5 x 8.25 x 5 in.
    7.5 in.
    21 in.
    10.5 cm
    3 mm, 5.65 mm
    0.5 cm
    1.65 lb.
    120 V
    CE
    Standard Electrophoresis Power Supply
    DNA
    0.5 to 6%
    Uses minimal consumables, no waste associated with precast gels
    Use with SmartGlow or SYBR Green Stain
    Electrophoresis
    12 or 22
    120 V
    Nucleic acid
    1 mm, 2 mm
    230 mL
    10 to 20 μL
    7.5 x 8.25 in.
    7.5 in.
    8.25 in.
    10 cm
    3 mm, 5.65 mm
    5 in.
    14 cm
    0.75 kg
    Videos
    SDS
    Documents

    Documents