missing translation for 'onlineSavingsMsg'
Learn More

Dynamic Diagnostics Reagent Tilter

Catalog No. 22899675
Encompass
Change view
Click to view available options
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22899675 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22899675 Supplier Dynamic Diagnostics Supplier No. 4000151
Only null left

Made from acrylic for safe and thorough dispensing

  • Tilt position makes dispensing reagents faster and safer
  • Can be placed on countertop or shelf
  • Dimensions: 10 x 11.75 x 4 in.

Specifications

Autoclavable Not Autoclavable
Color Clear
Material Acrylic
Sterility Non-sterile
For Use With (Equipment) Reagent Cubes
Width (English) 11.75 in.
Height (English) 4 in.
Depth (English) 10 in.