missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Reagent Starter Kits for 9300 and 9500 Photometers

Catalog No. 1540125
Change view
Click to view available options
Type:
Alkalinity, Total
Ammonia
Ammonia Conditioning Reagent
Calcium Hardness
Chloride
Chlorine-DPD Free, Comb, Total
Copper
Iron LR
Nitrate
Nitrite-N
pH (Phenol Red)
Potassium
Sulfate
Sulfide
Turbidity
Range:
0 to 0.5 mg/L
0 to 1 mg/L
0 to 10 mg/L
0 to 12 mg/L
0 to 20 mg/L
0 to 200 mg/L
0 to 5 mg/L
0 to 50 mg/L
0 to 500 mg/L
0 to 6 mg/L
5 to 400 mg/L
6.8 to 8.4 mg/L
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
1540125 Ammonia Conditioning Reagent 0 to 10 mg/L
1539915 Copper 0 to 5 mg/L
1539914 Turbidity 5 to 400 mg/L
153999 Chlorine-DPD Free, Comb, Total 0 to 6 mg/L
1539936 Potassium 0 to 12 mg/L
1539938 Sulfate 0 to 200 mg/L
153994 Ammonia 0 to 10 mg/L
153992 Alkalinity, Total 0 to 500 mg/L
1539939 Sulfide 0 to 0.5 mg/L
153997 Calcium Hardness 0 to 500 mg/L
1539928 Nitrate 0 to 20 mg/L
1539922 Iron LR 0 to 1 mg/L
153998 Chloride 0 to 50 mg/L
1539933 pH (Phenol Red) 6.8 to 8.4 mg/L
1539929 Nitrite-N 0 to 0.5 mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 1540125 Supplier YSI™ Supplier No. YAT1707
Only null left

For 9300 and 9500 Photometers.

  • Includes reagents sufficient for 50 tests

Specifications

Type Ammonia Conditioning Reagent
For Use With (Equipment) 9300 and 9500 Photometer
Range 0 to 10 mg/L