Learn More
YSI™ Reagent Starter Kits for 9300 and 9500 Photometers
For 9300 and 9500 Photometers.
Supplier: YSI™ YPM268
Description
- Includes reagents sufficient for 50 tests
Chemical Identifiers
| 9300 and 9500 Photometer |
Specifications
| Chloride | |
| Reagents, instructions, accessories | |
| 0 to 50 mg/L |
| 9300 and 9500 Photometer | |
| 50 Tests/Pk. |