missing translation for 'onlineSavingsMsg'
Learn More

Reagent Alcohol, ACS Reagent Grade Anhydrous, Absolute, Ricca Chemical

Catalog No. p-2915859
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
659016 500 mL Natural Poly Bottle
659032 1 L Natural Poly Bottle
659025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 659016 Supplier Ricca Chemical Company Supplier No. 659016
Only null left

Chemical Identifiers

Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS 64-17-5, 67-63-0, 67-56-1
CAS Min % 90.0, 4.0, 4.0
CAS Max % 91.0, 6.0, 5.0
Appearance Colorless Liquid
Quantity 500 mL
Packaging Natural Poly Bottle
Residue after Evaporation max. 0.001%
Water max. 0.5 %
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 76°C
Molecular Weight (g/mol) 46.069
Percent Purity 89.5 to 91.5%
Grade ACS
Chemical Name or Material Alcohol, Reagent , Anhyd., Absolute, Denatured alcohol
Show More Show Less
ShelfLife 36 months