missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyCount™ Stains

Catalog No. A49905
Encompass
Change view
Click to view available options
Type:
Viability Stain
Color:
Green/Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Color
A49905 Viability Stain Green/Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A49905 Supplier Invitrogen™ Supplier No. A49905

The ReadyCount stains are benchtop-stable fluorescent reagents for the Countess II FL and 3 FL automated cell counters. They are ready-to-use to enable fast and easy automated counting of nucleated cells or isolated nuclei.

ReadyCount stains are fluorescent cell stains specifically optimized for use on the Countess II FL and Countess 3 FL automated cell counters. These benchtop-stable reagents are ready-to-use nuclear stains that can be mixed 1:1 with cell samples. They are designed for fast and easy counting of nucleated cells and isolated nuclei, enabling increased accuracy using fluorescence-based count mode when counting mixed cell populations or samples containing cellular debris.

Features of the fluorescent ReadyCount stains:

  • Accurate—eliminates the error involved with creating stock solutions and dilution series
  • Flexible—optimized for use in both Countess II FL and Countess 3 FL automated cell counters equipped with DAPI, GFP, RFP, or Texas Red (TxR) light cubes
  • Fast—stains cells in < 5 minutes
  • Convenient—store on your benchtop, no freezing/thawing required, use by mixing 1:1 v/v with cell sample
  • Versatile—stain and count nucleated cells or isolated nuclei for RNA sequencing and gene expression applications in single-cell analysis workflows

ReadyCount stains work in three simple steps:

  • Mix 10 μL of cell sample with 10 μL of ReadyCount stain and incubate up to 5 minutes
  • Pipet 10 μL of the stained cell sample into a Countess chamber slide
  • Insert the slide into the Countess instrument

Obtain accurate cell counts

The fluorescent ReadyCount reagents allow for the specific staining of nucleated cells within a mixed population, thereby reducing complexity and improving count-to-count and sample-to-sample repeatability in automated cell counting when matched to the appropriate EVOS™ light cube.

Improve downstream single-cell analysis

Accurate viable cell counts are crucial when performing RNA sequencing and single-cell analysis. Staining with fluorescent ReadyCount stains and using the Countess™ 3 FL-based count mode enables detection of all fluorescent objects, resulting in more accurate counts and viability measurements for samples with debris or out of focus cells in brightfield. Characterization of isolated nuclei is also required for scATAC-Seq, a method for studying genome-wide chromatin accessibility. To accurately count isolated nuclei, it is recommended to use fluorescent ReadyCount stains instead of trypan blue. Trypan blue can lead to overcounting of nuclei due to debris staining.

The fluorescent ReadyCount stain options include:

  • ReadyCount Red Dead Cell Stain is a benchtop-stable red fluorescent stain optimized for staining dead nucleated cells in Countess II FL or Countess 3 FL automated cell counters. Traditional visible red filter sets such as the EVOS RFP or Texas Red light cubes are suitable for use with the ReadyCount Red Dead Cell Stain (propidium iodide).
  • ReadyCount Blue Nuclear Stain is a benchtop-stable blue fluorescent stain optimized for staining nucleated cells in Countess II FL or Countess 3 FL automated cell counters. Traditional DAPI filter sets such as the EVOS DAPI light cube are suitable for use with the ReadyCount Blue Nuclear Stain (Hoechst 33342).
  • ReadyCount Green/Red Dead Viability Stain is a benchtop-stable green and red fluorescent stain combination optimized for staining nucleated cells to measure viability in Countess II FL or Countess 3 FL automated cell counters. Traditional FITC or GFP filter sets such as the EVOS GFP light cube are suitable for exciting and detecting the green stain (acridine orange), while the EVOS Texas Red (TxR) light cube is suitable for the red cell stain (propidium iodide). Green denotes live cells while red represents dead cells. Because these dyes only stain nucleated cells, the ReadyCount Green/Red Dead Viability Stain is useful for obtaining accurate counts of viable cells for RNA sequencing and single-cell analysis.

Specifications

Type Viability Stain
Product Line ReadyCount
Compatibility Countess II FL >1.0.249; Countess 3 FL ≥1.0.296.782
For Use With (Equipment) Countess II FL, Countess 3 FL
Color Green/Red
Content And Storage ReadyCount™ Green/Red Viability Stain, 1 x 1 mL vial

Stable at room temperature.
Quantity 100 Assays
Shipping Condition Ambient
How can I use my Countess 3 Automated Cell Counter, for cell viability assays, and what are the steps?

Invitrogen ReadyCount Stains are pre-optimized for viability analysis with Countess 3 FL Automated Cell Counter. Simply mix 10 µL of cell sample with 10 µL of stain, pipet the solution into a Countess chamber or reusable slide and insert the slide into the instrument.

Which staining dyes can I use with my Countess 3 FL Automated Cell Counter?

EVOS light cubes, which enable fluorescent detection in the Countess 3 FL Automated Cell Counter, are compatible with a broad range of fluorescent dyes. Refer to our selection guide for an extensive list of compatible dyes. We also offer the ready-to-use ReadyCount Stains for easy viability staining.

What is the expiration date/warranty period for ReadyCount Green/Red Viability Stain (Cat. No. A49905)?

If an expiration date or warranty period is not posted on any public-facing document, the ReadyCount reagents have the standard warranty period of 1 year from the date of shipment.

What is the buffer/solution of the ReadyCount Green/Red Viability Stain (Cat. No. A49905)?

The formulation/identity of components in the solution of ReadyCount Green/Red Viability Stain (Cat. No. A49905) is proprietary.

What is the concentration of the Acridine Orange and Propidium iodide in ReadyCount Green/Red Viability Stain (Cat. No. A49905)?

The concentrations of Acridine Orange and Propidium iodide in ReadyCount Green/Red Viability Stain (Cat. No. A49905) are proprietary. If you need to use a defined concentration of Acridine Orange, we do offer this as a standalone product (Cat. Nos. A1301 and A3568). If you need to use a defined concentration of Propidium iodide, we do offer this as a standalone product (Cat. Nos. P3566, P1304MP, and P21493).

For Research Use Only. Not for use in diagnostic procedures.