missing translation for 'onlineSavingsMsg'
Learn More

Reactive Red 4, MP Biomedicals™

Catalog No. MP215201705
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215201705 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215201705 Supplier MP Biomedicals Inc Supplier No. 0215201705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibits phosphotransferase activity of yeast hexokinase. Reactive Red 4, MP Biomedicals protected the enzyme from inactivation by sugar and nucleotide substrates. This compound is a monochlorotriazine dye that contains polynuclear aromatic rings. Also kn

At the visible spectrum, Cibacron Brilliant Red 3B-A exhibits a ?max and ?min of 567nm and 501nm respectively. Studies suggest that the minimum structure required for this dye to bind hexokinase is 1-amino-8-naphthol-3,6-disulfonic acid.

Chemical Identifiers

CAS 17681-50-4
Molecular Formula C32H19ClN8Na4O14S4
Molecular Weight (g/mol) 995.195
InChI Key HMBFFLCYQUKBIW-UHFFFAOYSA-J
Synonym reactive red 4, cibacron brilliant red 3b-a, tetrasodium 5-benzamido-3-e-2-5-4-chloro-6-4-sulfonatophenyl amino-1,3,5-triazin-2-yl amino-2-sulfonatophenyl diazen-1-yl-4-hydroxynaphthalene-2,7-disulfonate
PubChem CID 44134921
IUPAC Name tetrasodium;5-benzamido-3-[[5-[[4-chloro-6-(4-sulfonatoanilino)-1,3,5-triazin-2-yl]amino]-2-sulfonatophenyl]hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)C(=O)NC2=C3C(=CC(=C2)S(=O)(=O)[O-])C=C(C(=NNC4=C(C=CC(=C4)NC5=NC(=NC(=N5)NC6=CC=C(C=C6)S(=O)(=O)[O-])Cl)S(=O)(=O)[O-])C3=O)S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 5 g
Solubility Information Soluble in water. Very slightly soluble in ethanol and methanol
Formula Weight 995.2
Physical Form Powder
Recommended Storage Room Temperature (15–30°C).