missing translation for 'onlineSavingsMsg'
Learn More

Reactive Blue #4 OR, Macron Fine Chemicals™

Catalog No. MK81321
Change view
Click to view available options
Quantity:
1kg
Packaging:
Poly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK81321 1kg Poly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK81321 Supplier Avantor Supplier No. 813203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13324-20-4
Molecular Formula C23H14Cl2N6O8S2
Molecular Weight (g/mol) 637.42
MDL Number MFCD00001218
InChI Key RTLULCVBFCRQKI-UHFFFAOYSA-N
PubChem CID 25863
IUPAC Name 1-amino-4-({3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfophenyl}amino)-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid
SMILES NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O

Specifications

Formula Weight 637.44
Packaging Poly
Quantity 1kg
Chemical Name or Material Reactive Blue #4, Procion Brilliant Blue MR