Learn More
Thermo Scientific™ Reacti-Vial™ Small Glass Reaction Vials
Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module.
Supplier:  Thermo Scientific™ TS13097
 View more versions of this product
Catalog No. PI13097
Only null left
Description
Description
Use Thermo Scientific™ Reacti-Vial™ Small Reaction Vials in the Reacti-Therm™ Heating and Stirring module. An internal cone makes small sample handling in these vials easy and convenient. The extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions. Reacti-Vials are available in a variety of sizes of clear and amber glass, and are supplied with Thermo Scientific™ Open-Top Screw Caps and PTFE/Rubber Discs.
Reacti-Vial™ Small Reaction Vials are ideal for:
  • Residue isolation
  • Derivative preparation
  • Maximum sample retrieval
  • Moisture protection
  • Sample storage
  • Precipitations
  • Centrifugations
  • Solvent evaporation
Specifications
Chemical Identifiers
Glass
Internal
Screw Cap (Open-top) with Septum
Certificate of Analysis (COA), Certificate of Conformance (COC)
PTFE/Rubber
Reacti-Vial
12 Pk.
Vial
Specifications
Amber
1 mL
Internal
1 mL
12 Pk.
Certificate of Analysis (COA), Certificate of Conformance (COC)
Glass
PTFE/Rubber
Reacti-Vial
Screw Cap (Open-top) with Septum
13-425
Vial
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.