missing translation for 'onlineSavingsMsg'
Learn More

RDX, SPEX CertiPrep™

Catalog No. S3251
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S3251 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S3251 Supplier SPEX CertiPrep Group LLC Supplier No. S3251
Only null left

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
RDX121-82-41000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 121-82-4
Molecular Formula C3H6N6O6
Molecular Weight (g/mol) 222.117
InChI Key XTFIVUDBNACUBN-UHFFFAOYSA-N
PubChem CID 8490
ChEBI CHEBI:24556
IUPAC Name 1,3,5-trinitro-1,3,5-triazinane
SMILES C1N(CN(CN1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]

Specifications

Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Min % 0.1
CAS Max % 0.1
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Acetonitrile:MeOH
Quantity 1 mL
Chemical Name or Material RDX
Hazard Category Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only