missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Color Coded Ziplock Biohazard Specimen Bags
Opaque colored specimen transport bags feature a reclosable seal with a tear away top for easy opening. Available in yellow, green, blue or red.
Description
- LDPE bags in 3 Wall style with 2 mil thickness
- Printed with black "SPECIMEN" and biohazard markings
- Red option also printed with "STAT"
- Color coded for easy specimen handling protocols
- Includes a paperwork pouch
Specifications
| Not Autoclavable | |
| Tear-away Zip-top | |
| Specimen transport | |
| LDPE | |
| 1000 | |
| 2 mil | |
| 9 in. | |
| 6 in. | |
| Transport Bag |
| 1.5 lb. | |
| Blue | |
| Biohazard print, paperwork pouch | |
| 0.279 mm | |
| 100 | |
| 9 x 6 in. (22.8 x 15.2cm) | |
| 22.8 cm | |
| 15.2 cm |