missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Color Coded Ziplock Biohazard Specimen Bags
Description
- LDPE bags in 3 Wall style with 2 mil thickness
- Printed with black "SPECIMEN" and biohazard markings
- Red option also printed with "STAT"
- Color coded for easy specimen handling protocols
- Includes a paperwork pouch
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Closure Type | Tear-away Zip-top |
| Color | Yellow |
| For Use With (Application) | Specimen transport |
| Includes | Biohazard print, paperwork pouch |
| Material | LDPE |
| No. per Case | 1000 |
| No. per Pack | 100 |
| Thickness (English) | 2 mil |
| Dimensions (L x W) | 9 x 6 in. (22.8 x 15.2cm) |
| Show More |