missing translation for 'onlineSavingsMsg'
Learn More

RD Plastics Three-wall Reclosable Biohazard Specimen Transport Bags

Resealable ziplock closure

$95.28 - $261.44

Specifications

Color Clear
For Use With (Application) Used for specimen transport
Thickness (Metric) 0.279 mm
Thickness (English) 2 mil
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Product Type Closure Type Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Product Type Closure Type Price Quantity  
01-800-20
View Documents
N/A 10 x 8 in. Biohazard Specimen Transport Bag Tear-away Top
Case of 1000 Each for $234.97
Only null left
 
23-700-205
View Documents
RD Plastics
RDB28
10 x 10 in. Biohazard Specimen Transport Bag Reclosable Flap
Case of 1000 Each for $261.44
Only null left
 
23-700-211
View Documents
RD Plastics
B122
9 x 6 in. Biohazard Specimen Transport Bag (Light Duty) Reclosable
Case of 1000 Each for $126.06
Only null left
 
23-700-209
View Documents
RD Plastics
RDB22
9 x 6 in. Biohazard Specimen Transport Bag Reclosable Flap
Case of 1000 Each for $95.28
Only null left
 
23-700-210
View Documents
N/A 10 x 8 in. Biohazard Specimen Transport Bag Reclosable Flap
Case of 1000 Each for $95.28
Only null left
 
Description

Description

  • Three-wall construction
  • Pouch has a flap cover to hold paperwork stays in place
  • With orange and black biohazard symbol and storage instructions

Specifications

Specifications

Clear
Used for specimen transport
0.279 mm
2 mil
Videos
SDS
Documents

Documents