missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Color Coded Ziplock Biohazard Specimen Bags
Color-coded 6 x 9 in. bags for easy identification. Available in green, red, pink or purple.
$151.03 - $180.48
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Closure Type | Zip Seal |
| For Use With (Application) | Specimen transport |
| Includes | Paperwork pouch |
| Material | LDPE |
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
23700202
|
RD Plastics
Q768 |
Green |
Case of 1000 Each for $151.03
|
|
|||||
|
23700204
|
RD Plastics
Q770 |
Pink |
Case of 1000 Each for $152.56
|
|
|||||
|
01800100
|
N/A | Purple |
Case of 1000 Each for $180.48
|
|
|||||
|
23700203
|
RD Plastics
Q769 |
Red |
Case of 1000 Each for $152.56
|
|
|||||
Description
Color coded biohazard/specimen transport bag with tear top and paperwork pouch- 2 mil clear reclosable bag with tear-away top for easy opening
- 3-wall construction for durable, reliable protection
- Biohazard symbol printed in black ink on colored block
Specifications
| Not Autoclavable | |
| Specimen transport | |
| LDPE | |
| 100 | |
| 9 in. |
| Zip Seal | |
| Paperwork pouch | |
| 1000 | |
| 22.86 x 15.25cm | |
| 6 in. |