missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RBS™ Solid Detergent

Catalog No. PI27968
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI27968 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI27968 Supplier Thermo Scientific™ Supplier No. 27968

A low-foaming, surface-active powdered detergent formula for use in laboratory automatic dishwashing machines.

Thermo Scientific™ RBS™ Solid Detergent is a low-foaming, surface-active powdered detergent formulated for use in automatic laboratory dishwashing machines.

Highlights:

  • For automatic laboratory washing machines Low-foaming
  • Rinses easily without leaving residues
  • Fully biodegradable

Specifications

Form Powder
Product Line RBS
Composition Anionic and Nonionic Detergent, 2% (v/v) Concentrate Solution, 0.2% (w/v) Solid Solution
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Quantity 5 kg
Volume (Metric) 5 kg
Which of your detergents are designed for use in automatic washers?

Only RBS Solid (Cat. No. 27968) is a non-foaming detergent that is suitable for automatic washers.

For Research Use Only. Not for use in diagnostic procedures.