missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RBS™ 35 Concentrate

Catalog No. PI27950
Change view
Click to view available options
Quantity:
1 kg
5 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI27950 1 kg
FER27852 20 kg
PI27952 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI27950 Supplier Thermo Scientific™ Supplier No. 27950
Only null left

A no-residue laboratory cleaning solution for labware utensils, surface materials and glassware, including bottles, beakers, pipets, lenses, mirrors.

Thermo Scientific RBS 35 Detergent Concentrate is a general cleaning agent for use on many laboratory surface materials and glassware, including lenses, mirrors, and reflectors.

Applications include the removal of:

  • Coomassie blue dye from test tubes and quartz cuvettes
  • Blood or serum residue from glass or plastic beakers and flasks
  • Colored dyes from porcelain mortar and pestles
  • HIV virus from glassware, tabletops and flooring
  • Radioactive isotopes from benchtops and instrument surfaces

The cleaning detergent is a mixture of anionic and nonionic surfactants. RBS™ 35 Detergent is strong enough to remove biological residues from plastic and glass, yet it is fully biodegradable and rinses away thoroughly to leave no residue. Use a 2% (v/v) solution for routine washing applications or soaking in a warm 20% solution to free particularly stubborn dry residues. Mix two parts 2% RBS Detergent solution with one part ethyl alcohol to remove greases and oils.

Complete removal of all impurities and contaminants from glassware is a necessity for most laboratory applications. For many years, caustic alcohol and chromic acid solutions were used for many substances that could not be cleaned from glassware by ordinary detergents. These strongly acidic or strongly basic cleaning products reduce the lifespan of glassware while exposing the user to potential harm. By contrast, Thermo Scientific RBS 35 and PCC-54 Products are mildly basic, biodegradable, surfactant concentrates containing anionic and nonionic detergents. The surfactants clean by penetrating between contaminating substances and the surfaces to be cleaned. Glass, plastics, quartz, porcelain and ferrous metal apparatus can be cleaned safely and effectively in a 2% solution (v/v) of the concentrates or a 0.2% solution (w/v) of the solids. RBS 35 and PCC-54 Cleaners remove both organic and inorganic materials and metal ions from laboratory equipment. The detergents rinse away completely, leaving no residue.

Note:

We do not sell RBS Products in Europe; EU customers can purchase our PCC-54 Products instead.

Specifications

Product Type RBS 35 Detergent Concentrate
Form Liquid
Product Line RBS
Composition 2% (v/v) Solution
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Quantity 1 kg
How can grease and oils be removed using the RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate??

To remove grease and oil, the detergents can be mixed with ethanol (e.g., two parts of a 2% solution to one part ethanol).

What materials are not compatible with the RBS 35 and PCC-54 detergents?

Because these detergents are alkaline, they may damage non-ferrous metals such as aluminum, zinc, magnesium and galvanized iron. Furthermore, neoprene rubber products should not be soaked in these detergents for extended periods of time.

What is the pH of the RBS 35 and PCC-54 detergents?

The pH of these detergents is slightly greater than 10.

What is the working concentration of the RBS 35 and PCC-54 detergents?

The working concentration for general-purpose applications is 2% for the concentrates and 0.3% for the automatic washer detergents. A 20% solution can be used for difficult cleaning situations.

What is the composition of the RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate?

RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate consist of ionic and nonionic surfactants compounded with strong water-softening cleaning powder components.

What is the difference between RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate?

RBS 35 Detergent Concentrate and PCC-54 Detergent Concentrate are similar products and may be used for the same applications. We do not sell RBS 35 Detergent Concentrate in Europe; Europe customers can purchase our PCC-54 35 Detergent Concentrate instead.


For Research Use Only. Not for use in diagnostic procedures.