Learn More
Eisco™ Raw Fluorite, Mineral Specimen - Hand Sample - Approx. 3 Inch
Fluorite Mineral Specimen is used in classroom identification lab exercises and for study of uncommon 4-plane cleavage pattern.
Supplier:  Eisco™ ESNG0014HAND
Catalog No. S02924
Only null left
Description
Description
  • Fluorite can be transparent or come in a variety of colors, such as purple, green and blue. Fluorite is a common mineral in both carbonate and hydrothermal rocks
  • Fluorite is perfect for hands on lessons in hardness, as well as the fickle method of identifying minerals by color. An excellent example for lessons in cleavage, as it is one of the few minerals that has four perfect planes of cleavage
  • Each specimen is selected and hand processed by an actual geologist, ensuring each sample exhibits defining features
  • Our rock and mineral samples are sourced from a variety of locations around the world and are processed in Victor, NY
  • Each specimen sample is packaged in a transparent bag, and is labeled and barcoded for easy identification
Specifications
Specifications
Mineral
3 in.
Product Suggestions
Product Suggestions