missing translation for 'onlineSavingsMsg'
Learn More

Rauwolscine hydrochloride, 99%

Catalog No. AAJ63198MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63198MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63198MC Supplier Thermo Scientific Chemicals Supplier No. J63198MC

CAS: 6211-32-1 | C21H27ClN2O3 | 390.908 g/mol

Standard alpha-2 adrenergic antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6211-32-1
Molecular Formula C21H27ClN2O3
Molecular Weight (g/mol) 390.908
MDL Number MFCD06668052
InChI Key PIPZGJSEDRMUAW-XQDOVXNESA-N
Synonym 17?-hydroxy-20?-yohimban-16?-carboxylic acid,methyl ester hydrochloride
PubChem CID 90479742
IUPAC Name methyl (1S,15S,18S,19S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
SMILES COC(=O)C1C(CCC2C1CC3C4=C(CCN3C2)C5=CC=CC=C5N4)O.Cl

Specifications

Color White
Quantity 100 mg
UN Number UN1544
Sensitivity Light sensitive
Formula Weight 390.91
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Rauwolscine hydrochloride
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber 228-279-6
RTECSNumber ZG1035000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only