missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ratchet Nut v3.0 Blue
Description
- Securing knob used to maintain the tube holder adapters on the central axis or 3 step assembly of the FastPrep-24™ instruments.
- Due to its high torque, it is particularly recommended for the heaviest adapters and especially for metal tube holders.
- Can be used on: FastPrep-24™ Classic or FastPrep-24™ 5G
Specifications
Specifications
| Product Type | Ratchet Nut |
| For Use With (Equipment) | FastPrep-24™ Classic, FastPrep-24™ 5G |
| Packaging Quantity | 1 Pk. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.