missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Rat Vegfa Recombinant Protein

Catalog No. 89967005
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89967005 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89967005 Supplier Abnova™ Supplier No. P4853
Only null left

Used for Func, SDS-PAGE

Sequence: MAPTTEGEQKAHEVVKFMDVYQRSYCRPIETLVDIFQEYPDEIEYIFKPSCVPLMRCAGCCNDEALECVPTSESNVTMQIMRIKPHQSQHIGEMSFLQHSRCECRPKKDRTKPEKHCEPCSERRKHLFVQDPQTCKCSCKNTDSRCKARQLELNERTCRCDKPRR

Specifications

Accession Number AAL07526.1
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 83785
Molecular Weight (g/mol) 38.8kDa
Name Vegfa (Rat) Recombinant Protein
Preparation Method Escherichia coli expression system
Quantity 10 μg
Immunogen MAPTTEGEQKAHEVVKFMDVYQRSYCRPIETLVDIFQEYPDEIEYIFKPSCVPLMRCAGCCNDEALECVPTSESNVTMQIMRIKPHQSQHIGEMSFLQHSRCECRPKKDRTKPEKHCEPCSERRKHLFVQDPQTCKCSCKNTDSRCKARQLELNERTCRCDKPRR
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias VEGF164/Vegf
Common Name Vegfa
Gene Symbol Vegfa
Biological Activity The activity is determined by the dose-dependent induced proliferation of HUVECs. The expected ED50 for this effect is 5.9-8.9ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less