Recombinant Protein;1MG;Rat ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-R52W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 27 - Gly 1130 (ITGA2) & Gln 21 - Asp 729 (ITGB1) (Accession # A0A0G2K470-1 (ITGA2) & P49134-1 (ITGB1)).;Rat ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA2 and ITGB1, has a calculated MW of 127.6 kDa (ITGA2) and 83.8 kDa (ITGB1). Subunit ITGA2 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus.