missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IgM Isotype Control (eBRM), PerCP-eFluor™ 710, eBioscience™

Catalog No. 46434180
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46434180 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46434180 Supplier Invitrogen™ Supplier No. 46434180
Only null left

Rat Monoclonal Isotype Control

The monoclonal rat IgM K is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control PerCP-eFluor 710 has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.

Specifications

Antigen Rat IgM
Applications Control, Flow Cytometry
Classification Monoclonal
Clone eBRM
Concentration 0.2 mg/mL
Conjugate PerCP-eFluor 710
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Isotype Control
Form Liquid
Isotype IgM κ
Show More Show Less