Learn More
Invitrogen™ Rat IgM Isotype Control (eBRM), Functional Grade, eBioscience™
Description
The monoclonal rat IgM, kappa is useful as an isotype control immunoglobulin. Applications Reported: Rat IgM Isotype Control Functional Grade Purified has been reported for use in flow cytometric analysis. Applications Tested: Rat IgM Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. It should be used at the same concentration as the experimental antibody. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Specifications
Specifications
| Antigen | Rat IgM |
| Applications | Control, Flow Cytometry, Functional Assay |
| Classification | Monoclonal |
| Clone | eBRM |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene Alias | Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202 |
| Host Species | Rat |
| Purification Method | Affinity chromatography |
| Show More |