missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IgG2b Isotype Control, Pacific Orange™

Catalog No. R2B30
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2B30 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2B30 Supplier Invitrogen™ Supplier No. R2B30

Rat Monoclonal Isotype Control

Rat IgG2b Isotype Control for Flow, Ctrl

Rat IgG2b is a protein-coding gene that encodes for the immunoglobulin G2b (IgG2b) subclass of rat antibodies. IgG2b is a major subclass of rat immunoglobulins that plays a crucial role in the immune response by neutralizing pathogens and promoting opsonization. Rat IgG2b is commonly used in research applications, such as in immunohistochemistry, flow cytometry, and Western blotting.

Specifications

Antigen Rat IgG2b
Applications Control, Flow Cytometry
Classification Monoclonal
Concentration 100 μg/mL
Conjugate Pacific Orange
Formulation PBS with 4mg/mL BSA and 0.1% sodium azide
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Rat
Immunogen Rat IgG2b
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 4°C, store in dark
Product Type Isotype Control
Form Liquid
Isotype IgG2b
Show More Show Less