missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rat IgG2a, PE, Clone: 2A3, Negative Control, EMD Millpore™

Catalog No. MABF1793MI
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
PE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABF1793MI 100 μg PE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1793MI Supplier MilliporeSigma™ Supplier No. MABF1793

Rat Monoclonal Antibody

Specifically detects IgG2a Clone: 2A3 in samples, and it is validated for Flow Cytometry

The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Specifications

Antigen IgG2a
Applications Flow Cytometry
Classification Monoclonal
Clone 2A3
Conjugate PE
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. This Monoclonal was purified from tissue culture supernatant via affinity chromatography.
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Test Specificity Rat IgG2a
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Isotype IgG2a
Show More Show Less

For Research Use Only