missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat GLUT4 (aa 493-509) Synthetic Peptide

Catalog No. ABC283
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC283 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC283 Supplier Invitrogen™ Supplier No. PEP283

Synthetic Peptide

Rat GLUT4 (residues 493-509) Synthetic Peptide, PEP-283, from Invitrogen.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).

Specifications

Gene ID (Entrez) 25139
Species Rat
Purification Method purified
Accession Number P19357
Common Name Rat GLUT4 (aa 493-509)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Slc2a4
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence E(493) - Q - E - V - K - P - S - T - E - L - E - Y - L - G - P - D - E – N - D (509)
Show More Show Less