missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat CSP alpha (aa 180-198) Synthetic Peptide

Catalog No. ABCB87
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABCB87 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABCB87 Supplier Invitrogen™ Supplier No. PEP287
Only null left

Synthetic Peptide

Rat CSP alpha (residues 180-198) Synthetic Peptide, PEP-287, from Invitrogen.

Cysteine string protein alpha (CSPalpha)/DNAJC5 is a synaptic vesicle protein that contains a DNA-J domain characteristic of Hsp40 chaperones. Cysteine-string proteins are membrane-bound proteins associated primarily with nerve endings and synaptic vesicles in the brain. These proteins play an important role in presynaptic function and may be involved in calcium-dependent neurotransmitter release. CSP proteins may act as presynaptic chaperones that assist in maintaining synaptic function.

Specifications

Gene ID (Entrez) 79130
Species Rat
Purification Method purified
Accession Number P60905
Common Name Rat CSP alpha (aa 180-198)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Dnajc5
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence E(180) - T - T - Q - L - T - A - D - S - H - P - S - Y - H - T - D - G - F - N(198)
Show More Show Less