missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat Calcium Sensing Receptor (aa 12-27) Synthetic Peptide

Catalog No. PEP009
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP009 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP009 Supplier Invitrogen™ Supplier No. PEP009
Only null left

Synthetic Peptide

Rat Calcium-Sensing Receptor (residues 12-27) Synthetic Peptide, PEP-009, from Invitrogen.

Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.

Specifications

Gene ID (Entrez) 24247
Species Rat
Purification Method purified
Accession Number P48442
Common Name Rat Calcium Sensing Receptor (aa 12-27)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Casr
Lyophilized Lyophilized
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence A - L - A - W - H - S - S - A - Y - G - P - D - Q - R - A - Q
Show More Show Less