Learn More
Ancare Corporation RAT CAGE POLYCARBONATE 10/CS
Catalog No. NC1386217
Encompass
Suitable for rats and guinea pigs. 10 12 x 19 x 8. Box only. Polycarbonate; clear, autoclavable to 250F (121C).
Catalog No. NC1386217 Supplier Ancare Corporation Supplier No. R20PC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for rats and guinea pigs. 10 12 x 19 x 8. Box only. Polycarbonate; clear, autoclavable to 250F (121C).